Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HUS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102763
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102763 Supplier Invitrogen™ Supplier No. PA5102763
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HUS1.

The protein encoded by this gene is a component of an evolutionarily conserved, genotoxin-activated checkpoint complex that is involved in the cell cycle arrest in response to DNA damage. This protein forms a heterotrimeric complex with checkpoint proteins RAD9 and RAD1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HUS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HUS1
Gene Accession No. O60921, Q8BQY8
Gene Alias Checkpoint protein HUS1; hHUS1; Hus1; HUS1 checkpoint clamp component; HUS1 checkpoint homolog; HUS1 checkpoint homolog (S. pombe); Hus1 homolog; Hus1 homolog (S. pombe); HUS1 protein; hus1+-like protein; mHUS1
Gene Symbols HUS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HUS1(Accession O60921), corresponding to amino acid residues E189-Q239.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15574, 3364, 498411
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.