Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HUS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576735
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The protein encoded by this gene is a component of an evolutionarily conserved, genotoxin-activated checkpoint complex that is involved in the cell cycle arrest in response to DNA damage. This protein forms a heterotrimeric complex with checkpoint proteins RAD9 and RAD1.
Specifications
HUS1 | |
Polyclonal | |
Unconjugated | |
HUS1 | |
Checkpoint protein HUS1; hHUS1; Hus1; HUS1 checkpoint clamp component; HUS1 checkpoint homolog; HUS1 checkpoint homolog (S. pombe); Hus1 homolog; Hus1 homolog (S. pombe); HUS1 protein; hus1+-like protein; mHUS1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15574, 3364, 498411 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.2 | |
O60921, Q8BQY8 | |
HUS1 | |
Recombinant full length Human HUS1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction