Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HUS1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566583
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566583 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566583 Supplier Invitrogen™ Supplier No. PA566583
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VEANLSGRMTL SIETEVVSIQ SYFKNLGNPP QSAVGVPENR DLESMVQVRV DNRKLLQFLE GQQIHPTTAL CNIWDNTLLQ LVLVQEDV Highest antigen sequence identity to the following orthologs - mouse 66%, rat 64%.

HUS1Bis most closely related to HUS1, a component of a cell cycle checkpoint protein complex involved in cell cycle arrest in response to DNA damage. HUS1B can interact with the check point protein RAD1 but not with RAD9. Overexpression of HUS1B has been shown to induce cell death, which suggests a related but distinct role of this protein, as compared to the HUS1. The protein encoded by this gene is most closely related to HUS1, a component of a cell cycle checkpoint protein complex involved in cell cycle arrest in response to DNA damage. This protein can interact with the check point protein RAD1 but not with RAD9. Overexpression of this protein has been shown to induce cell death, which suggests a related but distinct role of this protein, as compared to the HUS1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HUS1B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Hus1b
Gene Accession No. Q8NHY5
Gene Alias Checkpoint protein HUS1B; hHUS1B; HUS1 checkpoint clamp component B; HUS1 checkpoint homolog b; HUS1 checkpoint homolog b (S. pombe); Hus1 homolog b (S. pombe); Hus1b; MGC126746; MGC126748; mHUS1B; RP11-532F6.1
Gene Symbols Hus1b
Host Species Rabbit
Immunogen Recombinant Human HUS1B. Recombinant protein control fragment (Product #RP-107235).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135458
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.