Learn More
Invitrogen™ HUS1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA593237
Description
Immunogen sequence: MKFRAKITGK GCLELFIHVS GTVARLAKVC VLRVRPDSLC FGPAGSGGLH EARLWCEVRQ GAFQQFRMEG VSEDLDEIHL ELTAEHLSRA ARSAAGASSL KLQLTHKRRP SLTVAVELVS SLGRARSVVH DLPVRVLPRR VWRDCLPPSL RASDASIRLP RWRTLRSIVE RMANVGSHVL VEANLSGRMT LSIETEVVSI QSYFKNLGNP PQSAVGVPEN RDLESMVQVR VDNRKLLQFL EGQQIHPTTA LCNIWDNTLL QLVLVQEDVS LQYFIPAL; Positive Samples: LO2, Jurkat, Mouse spleen, Mouse thymus, Rat testis, Rat thymus.
HUS1Bis most closely related to HUS1, a component of a cell cycle checkpoint protein complex involved in cell cycle arrest in response to DNA damage. HUS1B can interact with the check point protein RAD1 but not with RAD9. Overexpression of HUS1B has been shown to induce cell death, which suggests a related but distinct role of this protein, as compared to the HUS1. The protein encoded by this gene is most closely related to HUS1, a component of a cell cycle checkpoint protein complex involved in cell cycle arrest in response to DNA damage. This protein can interact with the check point protein RAD1 but not with RAD9. Overexpression of this protein has been shown to induce cell death, which suggests a related but distinct role of this protein, as compared to the HUS1.
Specifications
HUS1B | |
Polyclonal | |
Unconjugated | |
Hus1b | |
Checkpoint protein HUS1B; hHUS1B; HUS1 checkpoint clamp component B; HUS1 checkpoint homolog b; HUS1 checkpoint homolog b (S. pombe); Hus1 homolog b (S. pombe); Hus1b; MGC126746; MGC126748; mHUS1B; RP11-532F6.1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
135458, 210554, 691382 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K572, Q8NHY5 | |
Hus1b | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-278 of human HUS1B (NP_6837622). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.