Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HUWE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100896
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100896 Supplier Invitrogen™ Supplier No. PA5100896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HUWE1.

SUFU encodes a component of the Sonic hedgehog (SHH; MIM 600725)/Patched (PTCH; MIM 601309) signaling pathway. Mutations in genes encoding components of this pathway are deleterious for normal development and are associated with cancer-predisposing syndromes (e.g., HPE3, MIM 142945; BCNS, MIM 109400; and GCPS, MIM 175700).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HUWE1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HUWE1
Gene Accession No. Q7TMY8, Q7Z6Z7
Gene Alias 5430439H10Rik; ARF-binding protein 1; ARF-BP1; AU041296; BJ-HCC-24 tumor antigen; C430014N20Rik; C80292; E3 ubiquitin-protein ligase HUWE1; E3Histone; Gm1718; HECT domain protein LASU1; HECT, UBA and WWE domain containing 1; HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase; HECT, UBA and WWE domain-containing protein 1; HectH9; HECT-type E3 ubiquitin transferase HUWE1; Homologous to E6AP carboxyl terminus homologous protein 9; HSPC272; HUWE1; Ib772; Kiaa0312; KIAA1578; Large structure of UREB1; LASU1; Mcl-1 ubiquitin ligase; Mcl-1 ubiquitin ligase E3; Mule; RP3-339A18.4; upstream regulatory element binding protein 1; upstream regulatory element-binding protein 1; Ureb1; URE-B1; URE-binding protein 1
Gene Symbols HUWE1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HUWE1(Accession Q7Z6Z7), corresponding to amino acid residues T2242-D2292.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10075, 59026
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.