Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HUWE1 Recombinant Rabbit Monoclonal Antibody (BLR129H)
GREENER_CHOICE

Catalog No. PIMA544447
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544447 100 μL
PIMA544448 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544447 Supplier Invitrogen™ Supplier No. MA544447
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.

SUFU encodes a component of the Sonic hedgehog (SHH; MIM 600725)/Patched (PTCH; MIM 601309) signaling pathway. Mutations in genes encoding components of this pathway are deleterious for normal development and are associated with cancer-predisposing syndromes (e.g., HPE3, MIM 142945; BCNS, MIM 109400; and GCPS, MIM 175700).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HUWE1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BLR129H
Concentration 10 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
Gene HUWE1
Gene Accession No. Q7TMY8, Q7Z6Z7
Gene Alias 5430439H10Rik; ARF-binding protein 1; ARF-BP1; AU041296; BJ-HCC-24 tumor antigen; C430014N20Rik; C80292; E3 ubiquitin-protein ligase HUWE1; E3Histone; Gm1718; HECT domain protein LASU1; HECT, UBA and WWE domain containing 1; HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase; HECT, UBA and WWE domain-containing protein 1; HectH9; HECT-type E3 ubiquitin transferase HUWE1; Homologous to E6AP carboxyl terminus homologous protein 9; HSPC272; HUWE1; Ib772; Kiaa0312; KIAA1578; Large structure of UREB1; LASU1; Mcl-1 ubiquitin ligase; Mcl-1 ubiquitin ligase E3; Mule; RP3-339A18.4; upstream regulatory element binding protein 1; upstream regulatory element-binding protein 1; Ureb1; URE-B1; URE-binding protein 1
Gene Symbols HUWE1
Host Species Rabbit
Immunogen Peptide representing a region between residue 2250 and 2300 of Human E3 ubiquitin-protein ligase HUWE1 (aka Lasu1, Ureb1).
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10075, 59026
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.