Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HVCN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558737 Supplier Invitrogen™ Supplier No. PA558737
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GIIISVKTRS ERQLLRLKQM NVQLAAKIQH LEFSCSEKEQ EIERLNKLLR QHGLLGEVN Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

HVCN1 (hydrogen voltage-gated channel 1), a voltage gated proton channel protein that is highly expressed in immune tissues, mediates the proton conductances required by phagocytic leukocytes for the oxidative burst that underlies microbial killing. HVCN1 moderates the voltage-dependent proton permeability of excitable membranes by allowing the flow of protons in accordance to their electrochemical gradient. HVCN1 is sensitive to zinc ions, and can be inhibited by them. HVCN1 may represent a new therapeutic target for B cell malignancies on continued signaling via the B cell antigen receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HVCN1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HVCN1
Gene Accession No. Q96D96
Gene Alias 0610039P13Rik; AI450555; BTS; HV1; Hvcn1; hydrogen voltage gated channel 1; hydrogen voltage-gated channel 1; mVSOP; RGD1310788; UNQ578/PRO1140; Voltage sensor domain-only protein; voltage-gated hydrogen channel 1; Vsop; Vsop.
Gene Symbols HVCN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HVCN1. Recombinant protein control fragment (Product #RP-91100).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84329
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.