Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HVEM/TNFRSF14 Antibody (028), DyLight 650

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289429C
Description
HVEM/TNFRSF14 Monoclonal specifically detects HVEM/TNFRSF14 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
HVEM/TNFRSF14 | |
Monoclonal | |
DyLight 650 | |
CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human HVEM/TNFRSF14 (NP_003811.2; Met1-Val202). | |
0.1 mL | |
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology | |
8764 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunofluorescence | |
28 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction