Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYAL1 Mouse anti-Human, Rat, Clone: 1B8B5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | HYAL1 |
---|---|
Clone | 1B8B5 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6979
![]() ![]() |
Proteintech Group Inc
665601IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a lysosomal hyaluronidase. Hyaluronidases intracellularly degrade hyaluro, one of the major glycosaminoglycans of the extracellular matrix. Hyaluro is thought to be involved in cell proliferation, migration and differentiation. This enzyme is active at an acidic pH and is the major hyaluronidase in plasma. Mutations in this gene are associated with mucopolysaccharidosis type IX, or hyaluronidase deficiency. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
HYAL1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HYAL 1, HYAL1, Hyaluronidase 1, hyaluronoglucosaminidase 1, LuCa 1, LUCA1, Lung carcinoma protein 1, NAT6 | |
HYAL1 | |
IgG1 | |
Protein G | |
Antibody |
1B8B5 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
Q12794, Q76HN1 | |
3373, 367166 | |
HYAL1 Fusion Protein Ag18183 | |
Primary | |
-20°C | |
HYAL1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title