Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HYAL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598451
Description
HYAL1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a lysosomal hyaluronidase. Hyaluronidases intracellularly degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan is thought to be involved in cell proliferation, migration and differentiation. This enzyme is active at an acidic pH and is the major hyaluronidase in plasma. Mutations in this gene are associated with mucopolysaccharidosis type IX, or hyaluronidase deficiency. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
HYAL1 | |
Polyclonal | |
Unconjugated | |
HYAL1 | |
Hya1; HYAL1; Hyal-1; Hyaluronidase; hyaluronidase 1; hyaluronidase-1; hyaluronoglucosaminidase 1; hyaluronoglucosaminidase-1; LUCA1; LuCa-1; Lung carcinoma protein 1; MPS9; NAT6; plasma hyaluronidase; tumor suppressor LUCA-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3373 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.56 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q12794 | |
HYAL1 | |
Recombinant Human Hyaluronidase-1 protein (136-435AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction