Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524223
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524223 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524223 Supplier Thermo Scientific Supplier No. PA524223
Add to Cart
Add to Cart

HYAL2 Polyclonal Antibody

HYAL2 Polyclonal Antibody for Western Blot, IHC (P), Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen HYAL2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene HYAL2
Gene Accession No. Q12891
Gene Alias AI256841; AU020858; AW555733; HYAL2; Hyal-2; Hyaluronidase; hyaluronidase 2; hyaluronidase-2; hyaluronoglucosaminidase 2; hyaluronoglucosaminidase-2; LUCA2; LuCa-2; lung carcinoma protein 2; lysosomal hyaluronidase; PH20 homolog; PH-20 homolog
Gene Symbols HYAL2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 385-412 amino acids from the C-terminal region of human HYAL2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8692
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.