Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HYAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557658
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557658 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557658 Supplier Invitrogen™ Supplier No. PA557658
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APPIFTGRPF VVAWDVPTQD CGPRLKVPLD LNAFDVQASP NEGFVNQNIT IFYRDRLGLY PRFDSAGRSV HGGVPQNVSL WAHRK Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.

This gene encodes a weak acid-active hyaluronidase. The encoded protein is similar in structure to other more active hyaluronidases. Hyaluronidases degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan and fragments of hyaluronan are thought to be involved in cell proliferation, migration and differentiation. Although it was previously thought to be a lysosomal hyaluronidase that is active at a pH below 4, the encoded protein is likely a GPI-anchored cell surface protein. This hyaluronidase serves as a receptor for the oncogenic virus Jaagsiekte sheep retrovirus. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. This gene encodes two alternatively spliced transcript variants which differ only in the 5' UTR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HYAL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HYAL2
Gene Accession No. Q12891
Gene Alias AI256841; AU020858; AW555733; HYAL2; Hyal-2; Hyaluronidase; hyaluronidase 2; hyaluronidase-2; hyaluronoglucosaminidase 2; hyaluronoglucosaminidase-2; LUCA2; LuCa-2; lung carcinoma protein 2; lysosomal hyaluronidase; PH20 homolog; PH-20 homolog
Gene Symbols HYAL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HYAL2. Recombinant protein control fragment (Product #RP-96270).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8692
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.