Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HYAL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561956
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561956 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561956 Supplier Invitrogen™ Supplier No. PA561956
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AAYQIHHSLR PGFAGPAVLD WEEWCPLWAG NWGRRRAYQA ASWAWAQQVF PDLDPQEQLY KAYTGFEQAA RALMEDTLRV AQALR Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 77%.

This gene encodes a member of the hyaluronidase family. Hyaluronidases are endoglycosidase enzymes that degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. The regulated turnover of hyaluronan plays a critical role in many biological processes including cell proliferation, migration and differentiation. The encoded protein may also play an important role in sperm function. This gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression, and the expression of specific transcript variants may be indicative of tumor status. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and some isoforms may lack hyaluronidase activity. This gene overlaps and is on the same strand as N-acetyltransferase 6 (GCN5-related), and some transcripts of each gene share a portion of the first exon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HYAL3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HYAL3
Gene Accession No. O43820
Gene Alias Hyal3; hyal-3; Hyaluronidase; hyaluronidase 3; hyaluronidase-3; hyaluronoglucosaminidase 3; hyaluronoglucosaminidase-3; Hyl3; LUCA14; LUCA3; LuCa-3; Lung carcinoma protein 3; Minna14
Gene Symbols HYAL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HYAL3. Recombinant protein control fragment (Product #RP-96473).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8372
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.