Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HYAL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556152
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556152 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556152 Supplier Invitrogen™ Supplier No. PA556152
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NNGRCIRKMW NAPSYLHLNP ASYHIEASED GEFTVKGKAS DTDLAVMADT FSCHCYQGYE GADCREIKTA DGCSGVSPSP GSLMTLCLLL LASYRSIQ Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 68%.

Endo-hyaluronidase that degrades hyaluronan to smaller oligosaccharide fragments. Has also chondroitin sulfate hydrolase activity, The best substrate being the galactosaminidic linkage in the sequence of a trisulfated tetrasaccharide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HYAL4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Hyal4
Gene Accession No. Q2M3T9
Gene Alias 4632428M18Rik; Chondroitin sulfate endo-beta-N-acetylgalactosaminidase; chondroitin sulfate hydrolase; CSHY; Hyal4; hyal-4; Hyaluronidase; hyaluronidase 4; hyaluronidase-4; hyaluronoglucosaminidase 4; hyaluronoglucosaminidase-4
Gene Symbols Hyal4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HYAL4. Recombinant protein control fragment (Product #RP-92670).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23553
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.