Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hyaluronan synthase 1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305924MFV610
Description
Hyaluronan synthase 1 Polyclonal antibody specifically detects Hyaluronan synthase 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Hyaluronan synthase 1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
HA synthase 1, HAS, huHAS1, hyaluronan synthase 1, hyaluronate synthase 1, hyaluronic acid synthase 1 | |
Produced in rabbits immunized with E. coli-derived Human Hyaluronan synthase 1 fragment. | |
0.1 mL | |
Angiogenesis, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction, Stem Cells | |
3036 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction