Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hyaluronic Acid, Bovine Vitreous Humor, PROMO, Thermo Scientific Chemicals

Catalog No. J60566MFPM
Click to view available options
Quantity:
50 mg
100 mg

Hyaluronic acid from bovine vitreous humor is a naturally derived glycosaminoglycan that shows an angiogenic activity that promotes wound healing processes.

General Description

  • Hyaluronic acid, derived from bovine vitreous humor, is a naturally derived, non-immunogenic, non-adhesive glycosaminoglycan that shows an angiogenic activity promoting wound-healing processes when degraded to small fragments

Application

  • It is considered a mucopolysaccharide composed of repeating dimeric units of glucuronic acid and N-acetyl glucosamine, which forms the core of complex proteoglycan aggregates found in the extracellular matrix
  • It has been used to study the activation of the MAPK/ERK pathway following infection of macrophages with Leishmania amazonensis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-61-9
Molecular Formula C28H44N2O23
Molecular Weight (g/mol) 776.65 g/mol
MDL Number MFCD00131348
InChI Key KIUKXJAPPMFGSW-YXBJCWEESA-N
Synonym hyaluronic acid, dsstox_cid_26750, dsstox_rid_81875, dsstox_gsid_46750, 4xi-alpha-d-xylo-hexopyranuronosyl-1->3-3xi-2-acetylamino-2-deoxy-beta-d-ribo-hexopyranosyl-1->4-5xi-beta-d-xylo-hexopyranuronosyl-1->3-2-acetylamino-2-deoxy-d-glucopyranose
PubChem CID 24728612
IUPAC Name (2S,4S,5R,6S)-6-[(2S,3R,5S,6R)-3-acetamido-2-[(3S,4R,5R,6R)-6-[(3R,4R,5S,6R)-3-acetamido-2,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-2-carboxy-4,5-dihydroxyoxan-3-yl]oxy-5-hydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES CC(=O)NC1C(C(C(OC1O)CO)O)OC2C(C(C(C(O2)C(=O)O)OC3C(C(C(C(O3)CO)O)OC4C(C(C(C(O4)C(=O)O)O)O)O)NC(=O)C)O)O

Specifications

Color White
Quantity 50 mg
Merck Index 14,4757
Solubility Information Soluble in water.
Physical Form Lyophilized Powder
Chemical Name or Material Hyaluronic acid
EINECSNumber 232-678-0
TSCA Yes
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.