Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hyaluronic acid Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA185561
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA185561 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA185561 Supplier Invitrogen™ Supplier No. PA185561
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

For direct ELISA, coat wells with 1% glutaraldehyde for 5 min at room temperature, followed by 3xPBS wash steps prior to coating with hyaluronic acid. Sheep anti hyaluronic acid antibody recognizes hyaluronic acid, a mucopolysaccharide which is an integral part of the gel-like substance of animal connective tissue that serves as a lubricating and protective agent.

Hyaluronic acid (HA), is the most abundant glycosaminoglycan (GAG) in mammalian tissue. It is present in high concentrations in connective tissue, such as skin, vitreous humor, cartilage, and umbilical cord, but the largest single reservoir is the synovial fluid (SF) of the diarthrodial joints, where concentrations of 0. 5-4 mg/mL are achieved. Hyaluronic acid, is the major hydrodynamic nonprotein component of joint SF. Its unique viscoelastic properties confer remarkable shock absorbing and lubricating abilities to SF, while its enormous macromolecular size and hydrophilicity serve to retain fluid in the joint cavity during articulation. HA restricts the entry of large plasma proteins and cells into SF but facilitates solute exchange between the synovial capillaries and cartilage and other joint tissues. In addition, HA can form a pericellular coat around cells, interact with proinflammatory mediators, and bind to cell receptors, such as cluster determinant (CD)44 and receptor for hyaluronate-mediated motility (RHAMM), where it modulates cell proliferation, migration, and gene expression. All these physicochemical and biologic properties of HA have been shown to be molecular weight (MW) dependent. The diverse physicochemical properties of HA arise from its unique macromolecular structure. The HA is an exceptionally long (3-30 μm) and unbranched nonsulfated GAG composed of repeating disaccharide units of N-acetylglucosamine, and glucuronic acid glycosidically linked through their respective 1-4 ring positions. Hydroxyl group oxygens at the glucuronyl-1 and glucosamine 3-positions are used for further polymerization of the HA disaccharide units to form chains that, when released from the cell plasma membrane, are of variable length and thus polydispersity. Despite the simplicity of the HA primary structure, this linear polyelectrolyte adopts complex conformations in solution, which engender it with diverse biologic properties. Within the joint cavity, HA molecules are predominately synthesized by the type B synovial cells. (PMID: 12219318). [National Center for Biotechnology Information. PubChem Compound Database; CID=24728612].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hyaluronic acid
Applications ELISA
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias C28H44N2O23
Host Species Sheep
Immunogen Hyaluronic acid from human umbilical cord.
Purification Method Protein G
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
How should I prepare my plates for a direct ELISA using the Hyaluronic acid Polyclonal Antibody (Cat. No. PA1-85561)?

For direct ELISA, we recommend coating wells with 1% glutaraldehyde for 5 min at room temperature, followed by 3X PBS wash steps before coating with the Hyaluronic acid Polyclonal Antibody (Cat. No. PA1-85561).

What is the molecular weight of the Hyaluronic acid Polyclonal Antibody (Cat. No. PA1-85561)?

The average molecular weight for the Hyaluronic acid Polyclonal Antibody (Cat. No. PA1-85561) is 150 kDa.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.