Learn More
Hyaluronic acid sodium salt, Streptococcus equi, 91%
Description
Bacterial hyaluronic acid (HA) is a natural non-sulfated high molecular weight glycosaminoglycan that may be used to study formulations of non-animal gels potentially useful for procedures such as soft tissue augmentation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Hyaluronic acid sodium salt is a component of the extracellular matrix that is potentially useful for soft tissue augmentation procedures
Application
• Hyaluronic acid sodium salt may be used in formulations of non-animal gels used in soft tissue augmentation studies
• It can be used in laboratory development of artificial synovial fluid
• It can also be used to study its role in oxidative stress and tumor progression
Specifications
Specifications
| Type | Hyaluronic Acid Sodium Salt |
| Physical Form | Powder |
| Color | White |
| Quantity | 500 mg |
| Merck Index | 14,4757 |
| Solubility Information | Soluble in water |
| Percent Purity | 91% |
| Chemical Name or Material | Hyaluronic acid sodium salt, Streptococcus equi |
Safety and Handling
| RTECSNumber | MT7250000 |
| TSCA | No |
| storageNote1 | -30° to -10°C |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.