Learn More
MilliporeSigma™ Supelco™ HybridSPE™ DPX SPE Tip, Bed Wt. 30 mg
Shop All MilliporeSigma Supelco ProductsDescription
HybridSPE technology is a simple and generic sample prep platform designed for the gross level removal of endogenous phospholipid interferences from biological plasma and serum prior to LC/MS or LC/MS/MS analysis. In this simple technique, biological plasma or serum is first subjected to protein precipitation via the addition and mixing of acidified acetonitrile. Precipitated proteins are then removed by centrifugation and the resulting supernatant is extracted using the HybridSPE DPX tip which acts as a chemical filter that specifically targets the removal of endogenous sample phospholipids. The phospholipid retention mechanism is based on a highly selective Lewis acid-base interaction between the proprietary zirconia ions functionally bonded to the HybridSPE stationary phase and the phosphate moiety consistent with all phospholipids. The resulting eluent is ready for immediate LC/MS or LC/MS/MS analysis.
MilliporeSigma Supelco offers an array of tips that are compatible with the most common liquid handling platforms and pipettors on the market: Tecan, Hamilton, INTEGRA, and a Universal format as well.
There are two versions of the DPX tips available for the HybridSPE-PL. The standard HybridSPE DPX 50 mg tips are designed for use with biological sample volumes between 100-300 muL, while the HybridSPE DPX 30 mg tips are designed for use with biological sample volumes between 30-100 muL.Guideline for 30 mg tips - plasma/serum sample: 30-100 muL; precipitating agent: 90-300 muL. Guideline for 50 mg tips - plasma/serum sample: 100-300 muL; precipitating agent: 300-900 muL.
- Minimal elution solvent volumes.
- Rapid extraction times (less than 3 minutes/sample).
- High extraction efficiencies.
- Easy to perform extractions.
- Lower costs.
- Higher throughput.
- Minimal training required.
- Environmentally friendly.
- Eliminates ion suppression effects often associated with phospholipids.
- Tips offered to fit many formats: Tecan, Hamilton, INTEGRA, and Universal liquid handling platforms and pipettors.
Specifications
Specifications
| Column Phase | DPX |
| Column Type | SPE |
| Bed Weight | 30 mg |
| For Use With (Application) | Food and Beverages |
| For Use With (Equipment) | Tecan |
| Product Line | HybridSPE |
| Type | SPE Tips |
| Volume (Metric) | 200 μL |
| Quantity | 96 Tips/Box |
HybridSPE is a registered trademark of Sigma-Aldrich Co. LLC
Your input is important to us. Please complete this form to provide feedback related to the content on this product.