Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HyClone™ 0.1 N NaOH, Cytiva

Catalog No. SH3088305 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
50 L
100 L
200 L
Packaging:
Single-use Bioprocessing Container
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SH3088305 50 L Single-use Bioprocessing Container
SH3088301 1 L Single-use Bioprocessing Container
SH3088303 10 L Single-use Bioprocessing Container
SH3088304 20 L Single-use Bioprocessing Container
SH3088306 100 L Single-use Bioprocessing Container
SH3088307 200 L Single-use Bioprocessing Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. SH3088305 Supplier Cytiva Supplier No. SH30883.05
Add to Cart
Add to Cart

Concentrated sodium hydroxide solutions for bioprocessing applications including pH adjustment and chromatography resin care.

HyClone™ NaOH solutions in single-use bags offer a convenient format to effectively modify pH and properly care for and clean chromatography resins.

  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.1 μm sterile filtered.
  • All unit sizes are filled in single-use, ready-to-use bags.
  • Raw materials are USP/NF and EP compendial grade.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

Color Clear, Colorless
Concentration 0.1 M (0.1 N)
Quantity 50 L
Packaging Single-use Bioprocessing Container
ShelfLife 12 Months
Recommended Storage 15°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.