Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HyClone™ 1 M NaCl, Cytiva

Catalog No. SH3114005 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
50 L
100 L
200 L
Packaging:
Single-use Bioprocessing Container
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SH3114005 100 L Single-use Bioprocessing Container
SH3114001 1 L Single-use Bioprocessing Container
SH3114002 10 L Single-use Bioprocessing Container
SH3114003 20 L Single-use Bioprocessing Container
SH3114004 50 L Single-use Bioprocessing Container
SH3114006 200 L Single-use Bioprocessing Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. SH3114005 Supplier Cytiva Supplier No. SH31140.05
Add to Cart
Add to Cart

Concentrated sodium chloride solutions for applications in upstream and downstream bioprocessing.

HyClone™ NaCl solutions in single-use bags offer a convenient format for many upstream and downstream bioprocessing applications.

  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.2 μm sterile filtered.
  • Provided in single-use bag containers.
  • Raw material is EP, USP, and JP grade.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color Clear, Colorless
Quantity 100 L
Concentration or Composition (by Analyte or Components) 1.0 M (1.0 N)
Packaging Single-use Bioprocessing Container
Product Line HyClone
ShelfLife 12 Months
Recommended Storage Room Temperature as defined by regional Pharmacopeia (15°C to 30°C in US; 15°C to 25°C in EU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.