Learn More
Description
ActiCHO P powdered cell culture medium is chemically defined, containing poloxamer 188, without insulin or L-glutamine.
- Hydrolysate-free and animal-derived component-free (ADCF). There are no proteins, peptides, or growth factors in ActiCHO P medium.
Specifications
Specifications
| Form | Powder |
| Sterility | Non-sterile |
| With Additives | Poloxamer 188 |
| Without Additives | No Glutamine, No Insulin |
| Quantity | 5 L |
| Content And Storage | 2 to 8°C, away from light and moisture |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.