Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva HyClone™ ActiCHO™ P Powdered Media
Click to view available options
Quantity:
1 L
5 L
Description
ActiCHO P powdered cell culture medium is chemically defined, containing poloxamer 188, without insulin or L-glutamine.
- Hydrolysate-free and animal-derived component-free (ADCF). There are no proteins, peptides, or growth factors in ActiCHO P medium.

Specifications
Specifications
Form | Powder |
Sterility | Non-sterile |
With Additives | Poloxamer 188 |
Without Additives | No Glutamine, No Insulin |
Quantity | 1 L |
Content And Storage | 2 to 8°C, away from light and moisture |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction