Learn More
Description
HyClone ActiSM medium is intended for use in combination with HyClone ActiPro medium and Cell Boost 7a and Cell Boost 7b supplements. This medium is part of the ActiPro system of media and supplements designed with adaptation and production in mind. HyClone ActiSM and the ActiPro system are suitable for general biomanufacturing with CHO cell lines such as CHO-GS, CHO-K1, CHO-DG44, and CHO-S.
- Chemically defined ADCF formulation optimized for high-yield protein production
- Does not contain hypoxanthine, thymidine, and glycine (HAT supplement)
- Recommended for first step when adapting cells cultured in other lean cell culture media
Specifications
Specifications
| Form | Powder |
| Sterility | Non-sterile |
| With Additives | Poloxamer 188 |
| Without Additives | No Glutamine |
| Quantity | 5 L |
| Content And Storage | 2 to 8°C, away from light and moisture |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.