Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ AdvanceSTEM™ Human Somatic Stem Cell Media

Catalog No. SH3088602 Shop All Cytiva Products
Change view
Click to view available options
Product Type:
Adipogenic Differentiation Medium
Chondrogenic Differentiation Medium
Osteogenic Differentiation Medium
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
SH3088602 Adipogenic Differentiation Medium
SH3088102 Osteogenic Differentiation Medium
SH3088902 Chondrogenic Differentiation Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3088602 Supplier Cytiva Supplier No. SH30886.02

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Research mesenchymal stem cells with HyClone™ AdvanceSTEM™ Human Somatic Stem Cell Media.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mesenchymal Stem Cells
Form Liquid
Product Type Adipogenic Differentiation Medium
Sterility Sterile-filtered
Quantity 450 mL
Content And Storage Store at 2° to 8°C away from light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.