Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Bovine Growth Serum

Catalog No. p-8013208
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated (25 to 40 doses)
None

A cost-effective alternative to Fetal Bovine Serum (FBS).

TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG
Content And Storage <–1°C
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing Irradiated (25 to 40 doses)
Tested For Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 10 mg/dL
Mycoplasma Detected Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.