Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Cell Boost 7a Supplement

Catalog No. p-7214286
Click to view available options
Quantity:
1 L
5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Supplements cell culture with amino acids, vitamins, salts, trace elements, and glucose. Manufactured to meet cGMP manufacturing standards and QC specifications.

Improve productivity and cell yield in fed-batch cultures by supplementing the cell culture medium to replace nutrients depleted during cell culture. HyClone Cell Boost 7a and Cell Boost 7b supplements are intended for use in combination with HyClone ActiPro ActiSM media as part of the ActiPro cell culture system formulated to provide high yields of recombinant proteins in processes when using Chinese hamster ovary (CHO) cell lines.

Cell Boost 7a is available as a convenient ready-to-use liquid feed solution or as a dry powder feed to be hydrated before use.

  • Designed to support modern manufacturing processes, Cell Boost 7a is chemically defined (CD), animal-derived component-free (ADCF), and does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol
  • Cell Boost 7a and 7b are concentrated cell culture supplements designed to be used together to enhance protein production
  • For convenience, this cell culture supplement is available as a ready-to-use liquid feed solution in addition to a dry powder format
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 to 8°C, away from light and moisture
Form Powder
Sterility Non-sterile
With Additives Poloxamer 188
Without Additives No Glutamine
Quantity 1 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.