Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva HyClone™ Characterized Fetal Bovine Serum (FBS), U.S. Origin
Click to view available options
Quantity:
500 mL
Processing:
HI, Irr - 25 to 40 doses
HI, Irr - 30 to 45 doses
Description
- Additional traceability testing of HyClone™ Fetal Bovine Serum is performed by a collaborator, Oritain
- Low in antibodies and high in growth factors
- Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
- Each lot is subjected to virus panel testing according to 9 CFR 113.53
- Characterized FBS is assayed for many components, including gamma globulin, glucose, and IgG
- Supports robust growth and division of cells, including embryonic stem cells
- Produced using true pool technology to minimize bottle-to-bottle variability
- Complete traceability is provided back to the original source, as certified by the International Serum Industry Association (ISIA)
- Sourced in the USA

Specifications
Specifications
Country of Origin | United States |
Form | Serum |
Product Type | Fetal Bovine Serum |
Quantity | 500 mL |
Sterility | Sterile |
Sterilization Method | Sequential Triple-filtered, 100 nm |
Grade | Characterized |
Processing | HI, Irr - 25 to 40 doses |
Tested For | Virus Panel Testing According to 9 CFR 113.53 |
Volume (Metric) | 500 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction