Learn More
Description
- Processed using a proprietary charcoal/dextran treatment that has been shown to reduce the levels of many hormones and growth factors
- This serum is assayed for a variety of components before and after treatment to ensure the efficacy of the treatment
- Charcoal/dextran treatment has been shown to be effective in reducing steroid levels for use of the serum in receptor studies or estrogen-related investigations
- Other studies indicate that charcoal/dextran treatments may be used to minimize lot-to-lot serum variability
- Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
- Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
Specifications
Specifications
| Additional Assays | 25 Dihydroxy Vitamin D, 17 Beta Estradiol, Albumin, Alkaline Phosphatase |
| Certifications/Compliance | EMEA Certified |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
| Country of Origin | United States |
| Quantity | 500 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential-filtered, 100 nm |
| Grade | Specialty filtered |
| Processing | Gamma Irr, 25 to 40 doses |
| Tested For | Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.