Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Fetal Bovine Serum (U.S.), Embryonic Stem (ES) Cell Screened

Catalog No. p-7058020
Click to view available options
Quantity:
500 mL
Processing:
Embryonic Stem Cell Screened
This item is not returnable. View return policy
This item is not returnable. View return policy

Augment growth of cells with U.S.-sourced HyClone™ Embryonic Stem (ES) Cell Screened Fetal Bovine Serum (FBS), filtered through serial 40nm pore-size rated filters and tested for endotoxin, hemoglobin, and extensive biochemical profile assays.

  • Filtered through sequential 40 nm (0.04μm) pore-size rated filters
  • Endotoxin: ≤ 10 EU/mL; Hemoglobin ≤ 10mg/dL
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays 1, 25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen, Calcium, Carotene, Chloride, Cholesterol, Copper, Creatinine, Folate, Gamma Globulin, Glucose, HDL, IgG, Inorganic Phosphorus, Insulin, Iron, Lactate Dehydrogenase, LDL, Magnesium, Niacin, Osmolality, Percent Saturation, pH, Phospholipids, Potassium, Progesterone, Retinol, Selenium, SGOT, SGPT, Sodium, TIBC, Tocopherol, Total Bilirubin, Total Protein, Triglycerides, Vitamin C
Certifications/Compliance EMEA Certified
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Country of Origin United States
Specialty Type ES cell screened
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 40 nm
Grade Defined, Specialty screened
Processing Embryonic Stem Cell Screened
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 10 mg/dL
Mycoplasma Detected Yes
USDA Safety Testing Included USDA Inspected
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.