Learn More
Description
Supplement cell culture growth with USDA-tested HyClone™ Fetal Bovine Serum (FBS), sourced from countries recognized as being free of BSE and FMD by the USDA.
- USDA safety-tested
- Filtered through three sequential 100 nm (0.1 μm) pore-size rated filters
Specifications
Specifications
| Additional Assays | Albumin |
| Certifications/Compliance | USDA Regulations |
| Content And Storage | <-1°C |
| Description | USDA Tested Fetal Bovine Serum, Central American Sourced |
| Country of Origin | Central America |
| Quantity | 500 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential-filtered, 100 nm |
| Source | Central America |
| Grade | Characterized |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.