Learn More
Description
Specifications
Specifications
| Additional Assays | Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Iron Saturation, Glucose, IgG |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. |
| Country of Origin | United States |
| Product Type | FBS Replacement Serum |
| Quantity | 500 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential-filtered, 100 nm |
| Processing | Irradiated (25 to 40 doses) |
| Tested For | Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
| Endotoxin Concentration | ≤ 10 EU/mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.