Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydantoin, 99%

Catalog No. AC168701000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

Catalog No. AC168701000

Supplier: Thermo Scientific Chemicals 168701000

Only null left
Add to Cart

CAS: 461-72-3 | C3H4N2O2 | 100.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-72-3
Molecular Formula C3H4N2O2
Molecular Weight (g/mol) 100.08
MDL Number MFCD00005259
InChI Key WJRBRSLFGCUECM-UHFFFAOYSA-N
Synonym hydantoin, 2,4-imidazolidinedione, glycolylurea, imidazole-2,4 3h,5h-dione, dantochlor, 2,4 3h,5h-imidazoledione, hydantoine, ccris 6532, 2-imidazolin-4 or 5-one, 2-hydroxy, 4h-imidazole-2,5-diol
PubChem CID 10006
ChEBI CHEBI:27612
IUPAC Name imidazolidine-2,4-dione
SMILES O=C1CNC(=O)N1

Specifications

Melting Point 220°C to 223°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 24, 242
Merck Index 15, 4799
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alkali hydroxide solutions,soluble in alkali,slightly soluble in ether
Formula Weight 100.08
Percent Purity 99%
Physical Form Fine Powder
Chemical Name or Material Hydantoin, 99%
Show More Show Less
EINECSNumber 207-313-3
RTECSNumber MT8210000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.