Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRA-POINT™ Coulometric Vessel Solution, CFC Free, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5391600
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    For Coulometric Karl Fischer Analysis

Specifications

Chemical Name or Material HYDRA-POINT™ Coulometric Vessel Solution
Quantity 500 mL
Flash Point 55.60°C
Name Note CFC Free
Packaging Narrow Mouth Amber Glass Bottle
Grade BAKER ANALYZED™ Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.