Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Composite 5 K, Reagent for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free), Honeywell™ Fluka™

Catalog No. 60046469
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
6 x 1 L

Specifications

Chemical Name or Material HYDRANAL™ - Composite 5 K
Quantity 6 x 500 mL
Name Note Reagent for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free)
Density ∼1.15 g/mL
Flash Point 99°C
CAS 111-90-0, 7553-56-2, 288-32-4, 693-98-1, 7446-09-5, 68007-08-9
CAS Min % ≥70.0000%, ≥5.0000%, ≥5.0000%, ≥1.0000%, ≥1.0000%, ≥1.0000%
CAS Max % <90.0000%, <10.0000%, <10.0000%, <5.0000%, <5.0000%, <5.0000%
Packaging Glass Bottle
Synonym DE Solvent, 2-(2-Ethoxyethoxy)ethanol, CARBITOL™, Diethylene glycol ethyl ether, Ethyldiglycol
Grade Karl Fischer
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.