Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYDRANAL™ - Coulomat AF 7, For coulometer AF7, Honeywell Fluka™
Click to view available options
Quantity:
1 L
6 x 1 L
Description
Advantages of Hydranal Coulometric Reagents
- Easy to use
- High accuracy for trace amounts of water
- Stable conditions of the titration vessel
- Broad product range
- Long shelf life (up to five years)

Specifications
Specifications
Chemical Name or Material | HYDRANAL™-Coulomat AF 7 |
CAS | 288-32-4, 7446-09-5, 67-56-1 |
Quantity | 1 L |
Flash Point | 11°C (51.8°F) |
Packaging | Glass bottle |
UN Number | UN1230 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction