Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Coulomat AF 7, For coulometer AF7, Honeywell Fluka™

Catalog No. p-7122072 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
6 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

for coulometer AF7

Advantages of Hydranal Coulometric Reagents

  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range
  • Long shelf life (up to five years)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Coulomat AF 7
CAS 288-32-4, 7446-09-5, 67-56-1
Quantity 1 L
Flash Point 11°C (51.8°F)
Packaging Glass bottle
UN Number UN1230
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.