Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYDRANAL™ - Coulomat CG-K, Reagent for coulometric KF titration in ketones (catholyte solution), Honeywell Fluka™

Click to view available options
Quantity:
1 Kit
10 x 5 mL Ampules
Specifications
Specifications
Chemical Name or Material | HYDRANAL™-Coulomat CG-K |
CAS | 7446-09-5, 288-32-4, 123-39-7 |
Quantity | 10 x 5 mL Ampules |
Flash Point | 64°C (147.2°F) |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction