Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Formamide dry, Solvent for KF titration, Honeywell Fluka™

Catalog No. 60090265 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L

Solvent for Karl Fischer titration

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Density 1.134g/mL at 25°C
CAS 75-12-7
Quantity 1 L
Flash Point 175°C (347°F)
Linear Formula HCONH2
Packaging Glass Bottle
UN Number NONH for all modes of transport
Beilstein 505995
Formula Weight 45.04g/mol
Chemical Name or Material HYDRANAL™-Formamide dry
Hazard Statement P201-P260-P280-P308 + P313
RTECSNumber LQ0525000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.