Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - LipoSolver MH, Reagent for volumetric one-component KF titration in non-polar substances, fats and oils (working medium), Honeywell Fluka™
BIOPHARMA

Catalog No. p-7122087 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
6 x 1 L
This item is not returnable. View return policy

Catalog No. 60-006-57

Supplier: Honeywell Chemicals 378566X1L

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Medium for volumetric one-component Karl Fischer titration in non-polar substances, fats and oils (methanol-hexanol-based)

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

Improved Composition

Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-LipoSolver MH
Density 0.80g/mL
CAS 67-56-1, 111-27-3
Quantity 1 L
Flash Point 11°C (51.8°F)
Packaging Glass bottle
MDL Number MFCD02687787
UN Number UN1230
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.