Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - NEXTGEN Solvent FI, Honeywell Fluka™

Catalog No. p-7203376 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
6 x 1 L

Catalog No. 60-049-485

Supplier: Honeywell Chemicals 344321L

Only null left
Add to Cart
Add to Cart

Medium for volumetric, two-component Karl Fischer titrations

  • Methanol-based
  • Free of Imidazole
  • Free of CMR Substances
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - NEXTGEN Solvent FI
CAS Min % ≥0.10%
Density 0.9 g/cm3 (at 20°C)
pH 5 to 6
Physical Form Liquid
CAS 147-85-3
Quantity 1 L
CAS Max % ≤5%
Flash Point 11°C
Packaging Glass Bottle
UN Number UN1230
Solubility Information Completely Miscible in Water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.