Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Solvent CM, Reagent for volumetric two-component KF titration in oils (working medium), Honeywell Fluka™

Catalog No. 6000665
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L

Specifications

Chemical Name or Material HYDRANAL™-Solvent CM
CAS 288-32-4, 67-56-1, 67-66-3
Density 1.26g/mL
Quantity 2.5 L
Flash Point 100°C (212°F)
MDL Number MFCD00147198
UN Number UN2810
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.