Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYDRANAL™ - Solvent CM, Reagent for volumetric two-component KF titration in oils (working medium), Honeywell Fluka™
Shop All Honeywell Research Chemicals Products

Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™-Solvent CM |
| CAS | 288-32-4, 67-56-1, 67-66-3 |
| Density | 1.26g/mL |
| Quantity | 1 L |
| Flash Point | 100°C (212°F) |
| MDL Number | MFCD00147198 |
| UN Number | UN2810 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction