Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYDRANAL™ - Solvent, Reagent for volumetric two-component KF titration (working medium), Honeywell Fluka™
Shop All Honeywell Research Chemicals Products

Click to view available options
Quantity:
1 L
2.5 L
Specifications
Specifications
Chemical Name or Material | HYDRANAL™-Solvent |
CAS | 67-56-1, 288-32-4, 7446-09-5 |
Density | 0.89g/mL |
Quantity | 2.5 L |
Flash Point | 11°C (51.8°F) |
MDL Number | MFCD00147197 |
UN Number | UN3286 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction