Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Solvent Oil, Reagent for volumetric two-component KF titration in oils (working medium), Honeywell Fluka™
BIOPHARMA

Catalog No. p-7122068
Click to view available options
Quantity:
1 L
6 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Medium for volumetric two-component Karl Fischer titration in non-polar substances, fats and oils (methanol-hexanol-based)

Advantages of Hydranal two-component reagents:

  • High titration speed
  • Ideal accuracy for small amounts of water
  • High buffer capacity
  • Exact and stable titer
  • E-type reagents: reduced toxicity compared to methanol
  • Long shelf life (three years for titrants, five years for solvents)

Improved Composition

In two-component reagents the Karl Fisher reactants are separated into two solutions: the titrant and the solvent. Hydranal-Titrant contains iodine dissolved in an alcohol with a precisely defined concentration. Hydranal-Solvent is an alcoholic solution of sulfur dioxide and imidazole. The alcohol is either methanol for standard reagents or ethanol for E-type reagents.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Solvent Oil
CAS 288-32-4, 67-56-1, 111-27-3
Density 0.83g/mL
Quantity 1 L
Flash Point 17°C (62.6°F)
MDL Number MFCD00147197
Packaging Glass bottle
UN Number UN1993BE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.