Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYDRANAL™-Solver (Crude) oil, Honeywell Fluka™
Shop All Honeywell Research Chemicals Products

Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Description
Advantages of Hydranal one-component reagents:
- Unlimited water capacity
- Convenient and easy to use
- The greatest flexibility in working media selection
- reduced toxicity compared to methanol based systems
- Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
- Long shelf life (three years)
Improved Composition
Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™-Solver (Crude) oil |
| CAS | 67-56-1, 7446-09-5, 67-66-3, 95-47-6, 288-32-4 |
| Density | 1.01g/mL |
| Quantity | 1 L |
| Flash Point | 6°C (42.8°F) |
| UN Number | UN1992 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction