Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™-Water-in-methanol 5.0, Honeywell Fluka™

Catalog No. 6000973 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-009-73 1 L
60-009-74 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-009-73 Supplier Honeywell Chemicals Supplier No. 348021L
Only null left
Add to Cart
Add to Cart

Titrating agent for volumetric back titration according to Karl Fischer, 5.00 mg/mL ± 0.05 mg/mL water in methanol

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name methanol
SMILES CO

Specifications

CAS 7732-18-5
Quantity 1 L
Flash Point 9°C (48.2°F)
UN Number UN1230
Beilstein 3662861
Formula Weight 32.04g/mol
Chemical Name or Material HYDRANAL™-Water-in-methanol 5.0
Hazard Statement P210-P260-P280-P301 + P310-P311
RTECSNumber PC1400000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.