Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYDRANAL™ - Water Standard KF-Oven 220°C-230°C, Standard for Karl Fischer oven check (water content ∼5.55%), Honeywell™ Fluka™
Click to view available options
Quantity:
10 g
Description
Advantages of HYDRANAL-Water Standards:
- Broad product range for volumetric and coulometric Karl Fischer applications, as well as the indirect oven application
- Manufactured according to ISO 9001 requirements
- Report of Analysis showing the exact water content and expanded uncertainty
Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222
- Long shelf life (up to five years)
- Convenient packaging
- Supplied with detailed instruction for use

Specifications
Specifications
CAS | 6100-05-6 |
Quantity | 10 g |
Packaging | Glass bottle |
UN Number | NONH for all modes of transport |
Chemical Name or Material | HYDRANAL™-Water Standard KF-Oven 220°C-230°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction