Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Hydrated-Sponge, Sold by AquaPhoenix Scientific
SCI_ED

Catalog No. p-7232127 Shop All AquaPhoenix Scientific Products
Click to view available options
Saturation Solution:
10 mL Neutralizing Buffer
10 mL letheen broth HS10LET

Includes

Case of 100

Uses simplified packaging to reduce waste and minimize handling and preparation

  • Ready-to-use, pre-hydrated sponges for easy and convenient environmental and food sample collection
  • Durable sponge withstands scrubbing
  • Sponge bag with write-on label for ease in labeling samples
  • Includes two decontaminated gloves for safety and convenience

Specifications

Product Type Hydrated Sponge
Length (English) 3 in.
Length (Metric) 7.6 cm
Width (English) 1.5 in.
Width (Metric) 4 cm
Packaging Quantity 100/Cs.
Saturation Solution 10 mL letheen broth HS10LET
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.