Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinylbenzyl trimethylammonium chloride, 97%

Catalog No. AC422561000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422561000 100 g Glass bottle
AC422560250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC422561000 Supplier Thermo Scientific Chemicals Supplier No. 422561000
Only null left
Add to Cart

CAS: 26616-35-3 | C12H18ClN | 211.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26616-35-3
Molecular Formula C12H18ClN
Molecular Weight (g/mol) 211.73
MDL Number MFCD00080690

Specifications

Melting Point 240°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH2CHC6H4CH2N(CH3)3Cl
Quantity 100 g
Formula Weight 211.73
Percent Purity 97%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Vinylbenzyl trimethylammonium chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
EINECSNumber 247-850-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.